Learn More
Chemglass Life Sciences Support Tubes, Stainless Steel, 1/2in OD X 36in Length
SDP
Catalog No. 50574104
Encompass_Preferred
O-Ring, Viton, #113. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200º C. For complete chemical resistance and fluid compatibility, see chart in the specifications tab at the bottom of this listing. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147. 13.94mm ID x 2.62mm Cross section.
This item is not returnable. View return policy
Catalog No. 50-574-104 Supplier Chemglass Life Sciences Supplier No. CG305105
O-Ring, Viton, #113. Viton o-ring is stocked in brown 75 durometer. They are suitable for acids, aliphatic hydrocarbons, oxidizing agents and all petroleum products. Viton has a low compression set and high temperature resistance. Viton is especially good for vacuum service and low gas permeability and has a useful temperature range of -30 to 200º C. For complete chemical resistance and fluid compatibility, see chart in the specifications tab at the bottom of this listing. FOR VITON O-RINGS USED WITH DURAN FLANGES, SEE CG-147. 13.94mm ID x 2.62mm Cross section.