Learn More
Beckman Coulter 4 mL, Open-Top Thinwall Ultra-Clear Tube, 11 x 60mm, 50/pk
Catalog No. NC9292622
Encompass_Preferred
Ultra-Clear Tubes, Size:11 x 60mm, Vol.: 4.0mL
Catalog No. NC9292622 Supplier Beckman Coulter Supplier No. 344062
Ultra-Clear Tubes, Size:11 x 60mm, Vol.: 4.0mL