Learn More
Beckman Coulter TUBES UC TN WALL 11X34MM (50)
Catalog No. NC9707088
Encompass_Preferred
Ultra-Clear Tubes, Size:11 x 34mm, Vol.: 2.2mL
Catalog No. NC9707088 Supplier Beckman Coulter Supplier No. 347356
Ultra-Clear Tubes, Size:11 x 34mm, Vol.: 2.2mL