Learn More
Bio Rad Laboratories TUBES WITH FLAT CAPS 0.2 ML
Catalog No. 509010029
Encompass
Tube; Bio-Rad; With frosted flat caps; Natural; With double-locking picture; Bio-Rad thermal cyclers with 96-, 48-, or 25-well reaction blocks accommodate 0.2mL tubes; PCR volumes of 5-125uL are recommended; 0.2mL; 1000
Catalog No. 50-901-0029 Supplier Bio Rad Laboratories Supplier No. TFI0201
Add to Cart
Add to Cart
Tube; Bio-Rad; With frosted flat caps; Natural; With double-locking picture; Bio-Rad thermal cyclers with 96-, 48-, or 25-well reaction blocks accommodate 0.2mL tubes; PCR volumes of 5-125uL are recommended; 0.2mL; 1000