Learn More
Mitegen Tubes, Wizard Classic 4, 48 formulations, 10 mL volumes
SDP
Catalog No. 501492095
Encompass_Preferred
The Wizard™ Classic line of random sparse matrix screens is designed to increase your probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).The Wizard Classic reagents are proven to be a highly effective starting point in the screening of biological macromolecules. The Wizard Classic formulations include a large range of crystallants, buffers, and salts covering a broad range of crystallization space at pH levels from pH 4.5 to pH 10.5.Choose from Wizard Classic 1, 2, 3, or 4 non-overlapping formulations in matrix blocks or tubes.Tube sets are 10 ml volumes per tube. All formulations are prepared with ASTM type-1 water and high-purity chemicals, 0.2 µm filtered in sterile packaging.
Catalog No. 50-149-2095 Supplier Mitegen Supplier No. 1009534
The Wizard™ Classic line of random sparse matrix screens is designed to increase your probability of producing crystals during the coarse screening phase when crystallizing biological macromolecules (proteins, nucleic acids, peptides, and combinations thereof).The Wizard Classic reagents are proven to be a highly effective starting point in the screening of biological macromolecules. The Wizard Classic formulations include a large range of crystallants, buffers, and salts covering a broad range of crystallization space at pH levels from pH 4.5 to pH 10.5.Choose from Wizard Classic 1, 2, 3, or 4 non-overlapping formulations in matrix blocks or tubes.Tube sets are 10 ml volumes per tube. All formulations are prepared with ASTM type-1 water and high-purity chemicals, 0.2 µm filtered in sterile packaging.