Learn More
Perkin Elmer US LLC Santoprene Two-Stop Peristaltic Pump Tubing - 1.14 mm I.D., Red/Red, Pkg. 12
Catalog No. 509052655
Encompass_Preferred
This Santoprene™ Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-905-2655 Supplier Perkin Elmer US LLC Supplier No. N0773119
This Santoprene™ Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.