Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBG TYG 2-STOP 1.42MM 12/PK
Catalog No. 502742876
Encompass
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-LFL , 1.42 mm ID, 15IN L; 12/PK, Avantor
Catalog No. 50-274-2876 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644734
Masterflex Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-LFL , 1.42 mm ID, 15IN L; 12/PK, Avantor