Learn More
Aviva Systems Biology Corp TUBG2 Antibody - middle region (ARP55345_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55345_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TUBG2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TUBG2. Alternate names: MGC131994ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-029