Learn More
ABclonal Technology TUBGCP6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A159211000UL
Encompass_Preferred
The protein encoded by this gene is part of a large multisubunit complex required for microtubule nucleation at the centrosome.
Catalog No. 50-001-05584