Learn More
SPEX CERTIPREP LLC 3P Turbidity QC Check Standards 10 NTU, 500 mL
Catalog No. NC0828738
Encompass_Preferred
(NSI by Zeptometrix) A formazin based CRM available as a single 500mL bottle or by the case of 4 x 500mL bottles. Stable and ready to use as received. Do not allow to freeze.
Catalog No. NC0828738 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI155
(NSI by Zeptometrix) A formazin based CRM available as a single 500mL bottle or by the case of 4 x 500mL bottles. Stable and ready to use as received. Do not allow to freeze.