Learn More
Qosina Corp Tygon® ND-100-80 Tubing, 80A Durometer; 0.02 inch ID; 0.06 inch OD (0.51 mm x 1.52 mm) x 500 ft coil (152.4 m)
Catalog No. NC3781427
Encompass
80A Durometer; 0.02 inch ID; 0.06 inch OD (0.51 mm x 1.52 mm) x 500 ft coil (152.4 m)
Catalog No. NC3781427 Supplier Qosina Corp Supplier No. T4102
80A Durometer; 0.02 inch ID; 0.06 inch OD (0.51 mm x 1.52 mm) x 500 ft coil (152.4 m)