Learn More
Thermo Scientific TUBING 0.10UMX190MM 1 FT LNGTH
Supplier:  Thermo Scientific 0010610499
Encompass
LC/MS Instrument Parts; Tubing, fused-silica, 0.10 ID x 0.19 OD, fused silica sample tube; Thermo Scientific; Replacement part for TSQ Quantum Triple Quadrupole LC/MS
Catalog No. NC9439557