Learn More
Helix Medical Inc Momentive Tubing | 0.30mm ID x 0.064mm OD x 0.15mm
Catalog No. NC1699009
Encompass_Preferred
All of our HelixMark Standard Silicone Tubing offerings are platinum-cured, and contain no additives, plasticizers, or peroxide components. The tubing is nonadherent and resists clotting, sticking, or encrustation. With the ability to operate at elevated temperatures, it will not oxidize or deteriorate. Rigorous testing, including expanded biocompatibility, provides quality assurance for the most demanding biomedical/pharmaceutical industry standards. HelixMark Standard Tubing can be sterilized with autoclave, ethylene oxide, or gamma radiation within the user’s validated cycles and exposure levels.
This item is not returnable. View return policy
Catalog No. NC1699009 Supplier Helix Medical Inc Supplier No. 6080501
All of our HelixMark Standard Silicone Tubing offerings are platinum-cured, and contain no additives, plasticizers, or peroxide components. The tubing is nonadherent and resists clotting, sticking, or encrustation. With the ability to operate at elevated temperatures, it will not oxidize or deteriorate. Rigorous testing, including expanded biocompatibility, provides quality assurance for the most demanding biomedical/pharmaceutical industry standards. HelixMark Standard Tubing can be sterilized with autoclave, ethylene oxide, or gamma radiation within the user’s validated cycles and exposure levels.