Learn More
IDEX Health & Science Upchurch TUBING .005 X .062 X 20CM
Catalog No. NC1919789
Encompass_Preferred
Idex Stainless-Steel Tubing 1/16 in. OD with Red Band, Diameter Inner: 0.125 mm, 1/16 in., Overall Length: 20 cm, 8 in., 316 Stainless Steel, Diameter Outer: 0.005 in., Pressure: 21,600 psi (1489 bar)
Catalog No. NC1919789 Supplier IDEX Health & Science Upchurch Supplier No. U154
Idex Stainless-Steel Tubing 1/16 in. OD with Red Band, Diameter Inner: 0.125 mm, 1/16 in., Overall Length: 20 cm, 8 in., 316 Stainless Steel, Diameter Outer: 0.005 in., Pressure: 21,600 psi (1489 bar)