Learn More
IDEX Health & Science Upchurch TUBING .010 X 20CM
Catalog No. NC1919770
Encompass_Preferred
Idex Stainless-Steel Tubing 1/16 in. OD with Blue Band, Diameter Inner: 0.25 mm, 1/16 in., Overall Length: 20 cm, 8 in., 316 Stainless Steel, Diameter Outer: 0.010 in., Pressure: 19,700 psi (1358 bar)
Catalog No. NC1919770 Supplier IDEX Health & Science Upchurch Supplier No. U113
Idex Stainless-Steel Tubing 1/16 in. OD with Blue Band, Diameter Inner: 0.25 mm, 1/16 in., Overall Length: 20 cm, 8 in., 316 Stainless Steel, Diameter Outer: 0.010 in., Pressure: 19,700 psi (1358 bar)