Learn More
IDEX Health & Science Upchurch TUBING .010 X 5CM
Catalog No. NC1919768
Encompass_Preferred
Idex Stainless-Steel Tubing 1/16 in. OD with Blue Band, Diameter Inner: 0.25 mm, 1/16 in., Overall Length: 5 cm, 2 in., 316 Stainless Steel, Diameter Outer: 0.010 in., Pressure: 19,700 psi (1358 bar)
Catalog No. NC1919768 Supplier IDEX Health & Science Upchurch Supplier No. U111
Idex Stainless-Steel Tubing 1/16 in. OD with Blue Band, Diameter Inner: 0.25 mm, 1/16 in., Overall Length: 5 cm, 2 in., 316 Stainless Steel, Diameter Outer: 0.010 in., Pressure: 19,700 psi (1358 bar)