Learn More
IDEX Health & Science Upchurch TUBING .040 X 5CM
Catalog No. NC1919782
Encompass_Preferred
Idex Stainless-Steel Tubing 1/16 in.OD 0.040 in.ID, Diameter Inner: 1.0 mm, 1/16 in., Overall Length: 5 cm, 2 in., 316 Stainless Steel, Diameter Outer: 0.040 in., Pressure: 8100 psi (558 bar)
Catalog No. NC1919782 Supplier IDEX Health & Science Upchurch Supplier No. U138
Idex Stainless-Steel Tubing 1/16 in.OD 0.040 in.ID, Diameter Inner: 1.0 mm, 1/16 in., Overall Length: 5 cm, 2 in., 316 Stainless Steel, Diameter Outer: 0.040 in., Pressure: 8100 psi (558 bar)