Learn More
Perkin Elmer US LLC Viton Two-Stop Peristaltic Pump Tubing - 1.14 mm I.D., Red/Red, Pkg. 12
Catalog No. 509047599
Encompass_Preferred
This Viton™ Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-904-7599 Supplier Perkin Elmer US LLC Supplier No. N0773115
This Viton™ Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.