Learn More
Accuflow Systems Tygon Tubing, Non-DEHP, 1/16 in ID, 1/8 in OD
Supplier:  Accuflow Systems ADF02002
Encompass_Preferred
Tygon non-DEHP surgical and hospital tubing is a clear, flexible medical-grade tubing formulated for blood-contact and general fluid-transfer applications. It provides a smooth inner bore for consistent flow, good chemical resistance, and soft walls that conform to fittings for tight routing in medical devices and laboratory setups.
  • Biocompatible, non-DEHP polymer suitable for medical applications.
  • Clear, flexible walls for easy routing and visual inspection.
  • Smooth inner bore to support consistent flow and minimal contamination.
  • Good chemical resistance to common medical and laboratory fluids.
  • Available in packaged lengths for bulk assembly and tubing runs.
Catalog No. NC2397377