Learn More
Trajan Scientific VICI Tubing, Polymeric, PTFE, 1/16" OD Tubing, 0.030" ID, 100 ft
Catalog No. 501090408
Encompass
VICI Valco Tubing, Polymeric, PTFE, 1/16" OD tuing, 0.030" ID, 100 ft. Made in the USA.
Catalog No. 50-109-0408 Supplier Trajan Scientific Supplier No. TTF130100
VICI Valco Tubing, Polymeric, PTFE, 1/16" OD tuing, 0.030" ID, 100 ft. Made in the USA.