Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Flexible Vinyl, 1/4"" ID x 3/8"" OD; 100 ft
Catalog No. NC1760572
Encompass
A good general-purpose tubing material, vinyl tubing (also known as PVC tubing) offers good chemical resistance. Its transparency allows easy visual identification of flow through the tube. This vinyl tubing's good abrasion resistance means you can use it in a laboratory, on a shop floor, or on a production bench. Vinyl tubing can be used within the temperature range of -45 to 180¿F (-43 to 82¿C).
Catalog No. NC1760572 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9648004
A good general-purpose tubing material, vinyl tubing (also known as PVC tubing) offers good chemical resistance. Its transparency allows easy visual identification of flow through the tube. This vinyl tubing's good abrasion resistance means you can use it in a laboratory, on a shop floor, or on a production bench. Vinyl tubing can be used within the temperature range of -45 to 180¿F (-43 to 82¿C).