Learn More
AVANTOR MASTERFLEX (VWR LLC) Bev-A-Line B-5 IV Tubing, 1/8""ID x 1/4""OD, 50 ft/pack
Catalog No. NC9214176
Encompass
Applications: Safe for food, beverage, drug, lab, and general use¿yet much more affordable than PTFE tubing. Safe for most alcohols; avoid use with strong solvents. Characteristics: Polyethylene liner with ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Heat-bondable. Translucent. Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements. Temperature range: ¿30 to 160¿F (¿51 to 71¿C) Sterilize: With ethylene oxide or dry heat.
Catalog No. NC9214176 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0649012
Applications: Safe for food, beverage, drug, lab, and general use¿yet much more affordable than PTFE tubing. Safe for most alcohols; avoid use with strong solvents. Characteristics: Polyethylene liner with ethyl vinyl acetate shell. No plasticizers. Nontoxic and nonhemolytic. Heat-bondable. Translucent. Certification: Resin is FDA-compliant (liner: 21 CFR 177.1520; shell 21 CFR 177.1350). Meets USP Class VI, USDA, and National Formulary requirements. Temperature range: ¿30 to 160¿F (¿51 to 71¿C) Sterilize: With ethylene oxide or dry heat.