Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC TUBING .125 OD .07ID 5FT
Catalog No. NC2250466
Encompass_Preferred
.125" OD x .07" ID x .0275 wall,hastalloy C-22 tubing, 5 ft
This item is not returnable. View return policy
Catalog No. NC2250466 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. 792705
.125" OD x .07" ID x .0275 wall,hastalloy C-22 tubing, 5 ft