Learn More
Accuflow Systems Saint-Gobain Tubing, 1/2 in ID, 3/4 in OD, 50 ft
Catalog No. 502365811
Encompass_Preferred
Tygon 2475 IB tubing is a crystal-clear, plasticizer-free thermoplastic tubing engineered for high-purity and medical fluid transfer applications. It minimizes extractables, offers low gas permeation compared with silicone, and meets USP Class VI biocompatibility standards. Typical uses include diagnostic reagent delivery, medical dispensing, cell and tissue culture transport, and cleanroom transfer lines.
  • Plasticizer-free thermoplastic construction minimizes extractables and leachables.
  • Crystal-clear wall enables visual inspection of the fluid path.
  • Smooth inner bore limits protein and particle retention.
  • Lower gas permeation than silicone, suitable for sensitive fluids.
  • 1/2 in ID, 3/4 in OD, 1/8 in wall, 50 ft length for long runs.
  • Designed for high-purity pressure applications and meets USP Class VI biocompatibility.
Catalog No. 50-236-5811 Supplier Accuflow Systems Supplier No. ACX00038
Tygon 2475 IB tubing is a crystal-clear, plasticizer-free thermoplastic tubing engineered for high-purity and medical fluid transfer applications. It minimizes extractables, offers low gas permeation compared with silicone, and meets USP Class VI biocompatibility standards. Typical uses include diagnostic reagent delivery, medical dispensing, cell and tissue culture transport, and cleanroom transfer lines.
  • Plasticizer-free thermoplastic construction minimizes extractables and leachables.
  • Crystal-clear wall enables visual inspection of the fluid path.
  • Smooth inner bore limits protein and particle retention.
  • Lower gas permeation than silicone, suitable for sensitive fluids.
  • 1/2 in ID, 3/4 in OD, 1/8 in wall, 50 ft length for long runs.
  • Designed for high-purity pressure applications and meets USP Class VI biocompatibility.