Learn More
Perkin Elmer US LLC Capillary Tubing for Mira Mist/Ari Mist - 0.44 in. O.D. x .018 in. I.D. x 0.5 m Long, Qty. 10
Catalog No. 509052408
Encompass_Preferred
This includes a 10 pack of clear Polyethylene tubing with attached BRC 3 fitting for sample lines.
Catalog No. 50-905-2408 Supplier Perkin Elmer US LLC Supplier No. N0777112
This includes a 10 pack of clear Polyethylene tubing with attached BRC 3 fitting for sample lines.