Learn More
Research Products International Corp Autoclavable Plastic Tubing, 2 ml Thick, 6 Inches Wide x 900 Feet Long
SDP
Catalog No. 50212682
Encompass
Create custom size bags for hybridization procedures or for autoclaving individual items. Ultra-clear 2 ml polypropylene tubing allows full visibility of bag contents. Tubing can be sealed quickly using an electric bag sealer.
Optional dispenser provides a convenient method for keeping tubing on a benchtop or can be wall mounted. Divider kit organizes various size rolls for convenient dispensing.
Catalog No. 50-212-682 Supplier Research Products International Corp Supplier No. 274560
Create custom size bags for hybridization procedures or for autoclaving individual items. Ultra-clear 2 ml polypropylene tubing allows full visibility of bag contents. Tubing can be sealed quickly using an electric bag sealer.
Optional dispenser provides a convenient method for keeping tubing on a benchtop or can be wall mounted. Divider kit organizes various size rolls for convenient dispensing.