Learn More
Cole-Parmer Idex 1524L Chromatography Tubing, Natural FEP, 3/16" OD x 0.125" ID x 50 ft L; 1/EA
Catalog No. NC1683567 Shop All Cole-Parmer Products
Encompass
FEP (fluorinated ethylene propylene) tubing is ideal choice for general, low pressure applications. Most FEP tubing is translucent, so you can see liquid flow.
Catalog No. NC1683567 Supplier Cole-Parmer Supplier No. 0201479
FEP (fluorinated ethylene propylene) tubing is ideal choice for general, low pressure applications. Most FEP tubing is translucent, so you can see liquid flow.