Learn More
AVANTOR MASTERFLEX (VWR LLC) Lab Tubing, Non-DEHP, 3/32""ID X 5/32""OD, 50 FT/Pk
Catalog No. NC0782179
Encompass
E-3603 Non-DEHP Lab Tubing, 3/32" ID x 5/32" OD; 50 ft/Pk, Formulation: Tygon E-3603Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments.Characteristics: Non-DEHP, phthalate free. Long life, chemical resistant, nonoxidizing; able to bend to sharp radius curves and easily works with fittings. Transparent.Certifications: FDA compliant; meets USP Class VI and NSF 51 requirements.Temperature range: ¿51 to 165¿F (¿46 to 74¿C)
Catalog No. NC0782179 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640773
E-3603 Non-DEHP Lab Tubing, 3/32" ID x 5/32" OD; 50 ft/Pk, Formulation: Tygon E-3603Applications: General laboratory, food and beverage, biopharmaceutical, analytical instruments.Characteristics: Non-DEHP, phthalate free. Long life, chemical resistant, nonoxidizing; able to bend to sharp radius curves and easily works with fittings. Transparent.Certifications: FDA compliant; meets USP Class VI and NSF 51 requirements.Temperature range: ¿51 to 165¿F (¿46 to 74¿C)