Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Norprene, 3/8"" ID x 1/2"" OD; 50 ft
Catalog No. NC1555041
Encompass
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275¿F (-60 to 135¿C). Can be sterilized with autoclave.
Catalog No. NC1555041 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641010
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275¿F (-60 to 135¿C). Can be sterilized with autoclave.