Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Platinum-Cured Silicone, 3/8"" ID x 5/8"" OD; 300 ft
Catalog No. NC1075509
Encompass
Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing. Platinum-cured silicone spools are sold in 300 ft or 500 ft spools.
Catalog No. NC1075509 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9890610
Add to Cart
Add to Cart
Request a Quote
Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing. Platinum-cured silicone spools are sold in 300 ft or 500 ft spools.