Learn More
Grainger TUBING,3/8X1/4"ID,NORPRENE,50'
Catalog No. NC2024292
Encompass_Preferred
50 ft. Norprene(R), A-60-G Thermoplastic Elastomer Tubing, 3/8 in Outside Dia., 1/4 in Inside Dia
Catalog No. NC2024292 Supplier Grainger Supplier No. 2DCD3
50 ft. Norprene(R), A-60-G Thermoplastic Elastomer Tubing, 3/8 in Outside Dia., 1/4 in Inside Dia