Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 3/8 in I.D., 5/8 in O.D.
Catalog No. 502365834
Encompass_Preferred
High-purity, textile-reinforced thermoplastic tubing designed for pressurized fluid transfer. The tubing features a smooth inner bore to preserve product integrity and is suitable for laboratory, pharmaceutical, and industrial fluid-handling applications.
  • High-purity thermoplastic construction.
  • Textile reinforcement for increased pressure resistance.
  • Inner diameter 3/8 in (9.5 mm).
  • Outer diameter 5/8 in (15.9 mm).
  • Wall thickness 1/8 in (3.2 mm).
  • Length 50 ft (15.2 m).
  • Smooth inner bore to minimize fluid interaction.
Catalog No. 50-236-5834 Supplier Accuflow Systems Supplier No. ACX00029
High-purity, textile-reinforced thermoplastic tubing designed for pressurized fluid transfer. The tubing features a smooth inner bore to preserve product integrity and is suitable for laboratory, pharmaceutical, and industrial fluid-handling applications.
  • High-purity thermoplastic construction.
  • Textile reinforcement for increased pressure resistance.
  • Inner diameter 3/8 in (9.5 mm).
  • Outer diameter 5/8 in (15.9 mm).
  • Wall thickness 1/8 in (3.2 mm).
  • Length 50 ft (15.2 m).
  • Smooth inner bore to minimize fluid interaction.