Learn More
McMaster-Carr Extreme-Temperature Teflon PTFE Semi-Clear Tubing for Chemicals, 3/16" ID, 1/4" OD
Catalog No. NC1860151
Encompass
Run acids, coolant, and other harsh chemicals through this PTFE tubing over a wide range of temperatures. It has a smooth interior for unrestricted flow and easy cleaning. Tubing is semi-flexible, so it is good for gradual bends. Not for use in medical applications.
Catalog No. NC1860151 Supplier McMaster-Carr Supplier No. 5239K12
Run acids, coolant, and other harsh chemicals through this PTFE tubing over a wide range of temperatures. It has a smooth interior for unrestricted flow and easy cleaning. Tubing is semi-flexible, so it is good for gradual bends. Not for use in medical applications.