Learn More
Waters Corp Waters Tubing Assembly, Accumulator to Lower Transducer, HPLC System, Stainless Steel
Catalog No. 50793826 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This spare part is a tubing assembly designed for connecting the accumulator to the lower transducer in an HPLC system. It is constructed from stainless steel to ensure durability and compatibility with various laboratory applications.
  • Tubing for connecting the accumulator to the lower transducer.
  • Suitable for HPLC systems.
  • Constructed from stainless steel.
  • Provided as a single unit per package.
  • Designed for replacement as required, ensuring optimal system performance.
Catalog No. 50-793-826 Supplier Waters Corp Supplier No. WAT270971
This spare part is a tubing assembly designed for connecting the accumulator to the lower transducer in an HPLC system. It is constructed from stainless steel to ensure durability and compatibility with various laboratory applications.
  • Tubing for connecting the accumulator to the lower transducer.
  • Suitable for HPLC systems.
  • Constructed from stainless steel.
  • Provided as a single unit per package.
  • Designed for replacement as required, ensuring optimal system performance.