Learn More
Waters Corp Waters Tubing Assembly, HPLC, Stainless Steel, Accumulator to Top Transducer
Catalog No. 50793825 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed to connect the accumulator to the top transducer in a system. It is made from stainless steel and is suitable for use in HPLC systems.
  • Designed as a spare part.
  • Constructed from stainless steel.
  • Compatible with HPLC systems.
  • Connects the accumulator to the top transducer.
Catalog No. 50-793-825 Supplier Waters Corp Supplier No. WAT270970
This tubing assembly is designed to connect the accumulator to the top transducer in a system. It is made from stainless steel and is suitable for use in HPLC systems.
  • Designed as a spare part.
  • Constructed from stainless steel.
  • Compatible with HPLC systems.
  • Connects the accumulator to the top transducer.