Learn More
Perkin Elmer US LLC 1⁄4 in. (6.4 mm) External Screw Thread Adapter with 1.8 mm O.D. Nipple, Type K
Catalog No. 509042489
Encompass_Preferred
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This connector is included in the Hydride Accessory Kit for Avio 200/500 (N0810433). Type K in instrument manual diagrams.
Catalog No. 50-904-2489 Supplier Perkin Elmer US LLC Supplier No. B0507918
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. This connector is included in the Hydride Accessory Kit for Avio 200/500 (N0810433). Type K in instrument manual diagrams.