Learn More
Perkin Elmer US LLC 1⁄4 in. (6.4 mm) External Screw Thread Adapter with 0.7 mm O.D. Pt/lr Capillary Nipple, Type N
Catalog No. 509042494
Encompass_Preferred
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. Type N in instrument manual diagrams.
Catalog No. 50-904-2494 Supplier Perkin Elmer US LLC Supplier No. B0507949
Tubing Adapters and Connectors provide a means to connect the component shown to the screw fittings of the PTFE tubing assemblies or to connect the tubing components described. Type N in instrument manual diagrams.