Learn More
Sigma Aldrich Fine Chemicals Biosciences Tubing adapter with Torion1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5319011EA
Encompass
PTfE stopcocks are threaded for additional safety and have a 2 mm bore for 3/16 in. tubing connection and 4 mm bore for 5/16 in. and 1/2 in. tubing connections. 5/16in. O.D fits a 8 Torion and 1/2in. O.D. fits a 14 Torion.
Catalog No. 50-290-5088