Learn More
Waters Corp Tubing Assembly, Accumulator ‘B’ to Transducer, MP35N
Catalog No. 502392655 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for connecting an accumulator ‘B’ to a transducer. It is made from MP35N material, ensuring durability and compatibility in its intended application.
  • Designed for connecting accumulator ‘B’ to a transducer.
  • Constructed from MP35N material.
  • Functions as a spare parts kit.
Catalog No. 50-239-2655 Supplier Waters Corp Supplier No. 700009849
This tubing assembly is designed for connecting an accumulator ‘B’ to a transducer. It is made from MP35N material, ensuring durability and compatibility in its intended application.
  • Designed for connecting accumulator ‘B’ to a transducer.
  • Constructed from MP35N material.
  • Functions as a spare parts kit.