Learn More
Waters Corp Waters Tubing Assembly, ACQUITY UPLC I-Class, Column to PDA, Stainless Steel, 10.5 in.
Catalog No. 502039244 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use with ACQUITY UPLC I-Class systems. It connects the column to a PDA detector and is constructed from stainless steel with a length of 10.5 in. It is suitable for applications where only one detector is being utilized.
  • Connects an I-Class system to a PDA detector
  • Designed for use when only one detector is being utilized
Catalog No. 50-203-9244 Supplier Waters Corp Supplier No. 205000895
This tubing assembly is designed for use with ACQUITY UPLC I-Class systems. It connects the column to a PDA detector and is constructed from stainless steel with a length of 10.5 in. It is suitable for applications where only one detector is being utilized.
  • Connects an I-Class system to a PDA detector
  • Designed for use when only one detector is being utilized