Learn More
Waters Corp Tubing Assembly, .007 in. ID, Stainless Steel
Catalog No. 502392096 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is designed for APH to Column connections. It features a .007 in. ID and is constructed from stainless steel, making it suitable for use in CH-30A and ideal for higher flow applications.
  • Replacement tubing assembly
  • Designed for APH to Column connection
  • Features a .007 in. ID
  • Constructed from stainless steel
  • Suitable for use in CH-30A
  • Ideal for higher flow applications
Catalog No. 50-239-2096 Supplier Waters Corp Supplier No. 700010698
This replacement tubing assembly is designed for APH to Column connections. It features a .007 in. ID and is constructed from stainless steel, making it suitable for use in CH-30A and ideal for higher flow applications.
  • Replacement tubing assembly
  • Designed for APH to Column connection
  • Features a .007 in. ID
  • Constructed from stainless steel
  • Suitable for use in CH-30A
  • Ideal for higher flow applications