Learn More
Waters Corp Tubing Assembly, Bio Pump Outlet, 0.062 in. OD, 0.010 in. ID, 14 in. Length
Catalog No. 502390724 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for precise fluidics connections in various UPLC systems, specifically for the bio pump outlet. It ensures reliable performance and compatibility with several system configurations.
  • Designed for bio pump outlet applications.
  • Compatible with specific UPLC system configurations including QSM, H-Class Bio, and Premier types.
  • Features a 0.062 in. OD, 0.010 in. ID, and 14 in. length.
  • Connects pump outlet (mixer) to injection valve port 5.
  • Suitable for UPLC system applications.
  • Replacement as required.
Catalog No. 50-239-0724 Supplier Waters Corp Supplier No. 430002473
This tubing assembly is designed for precise fluidics connections in various UPLC systems, specifically for the bio pump outlet. It ensures reliable performance and compatibility with several system configurations.
  • Designed for bio pump outlet applications.
  • Compatible with specific UPLC system configurations including QSM, H-Class Bio, and Premier types.
  • Features a 0.062 in. OD, 0.010 in. ID, and 14 in. length.
  • Connects pump outlet (mixer) to injection valve port 5.
  • Suitable for UPLC system applications.
  • Replacement as required.