Learn More
Waters Corp Waters Tubing Assembly, Capillary, Fused Silica, 75 μm ID, 10 in.
Catalog No. 502039750 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing assembly is designed for use in UPLC M-Class with HDX Technology systems. It features a fused silica tube with fittings and is specifically engineered for plumbing connections, facilitating critical fluid transfer within the system.
  • Features a fused silica tube with fitting.
  • Dimensions of 75 μm ID × 10 in.
  • Used for plumbing connections in manual HDX systems.
  • Connects to VanGuard trap column.
  • Connects to trap valve ports.
  • Connects to HDX fluid block.
  • Connects to analytical column inlet.
Catalog No. 50-203-9750 Supplier Waters Corp Supplier No. 700009871
This capillary tubing assembly is designed for use in UPLC M-Class with HDX Technology systems. It features a fused silica tube with fittings and is specifically engineered for plumbing connections, facilitating critical fluid transfer within the system.
  • Features a fused silica tube with fitting.
  • Dimensions of 75 μm ID × 10 in.
  • Used for plumbing connections in manual HDX systems.
  • Connects to VanGuard trap column.
  • Connects to trap valve ports.
  • Connects to HDX fluid block.
  • Connects to analytical column inlet.