Learn More
Waters Corp Tubing Assembly, Capillary, Column Inlet, 300 μm
Catalog No. 502392969 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This capillary tubing assembly functions as a column inlet and is designed for compatibility with specific nanoACQUITY system configurations, ensuring reliable performance in high-pressure applications.
  • 300 μm column inlet capillary tube.
  • Fits 5000 psi and 10,000 psi nanoACQUITY systems.
  • 1 unit per package.
Catalog No. 50-239-2969 Supplier Waters Corp Supplier No. 700002754
This capillary tubing assembly functions as a column inlet and is designed for compatibility with specific nanoACQUITY system configurations, ensuring reliable performance in high-pressure applications.
  • 300 μm column inlet capillary tube.
  • Fits 5000 psi and 10,000 psi nanoACQUITY systems.
  • 1 unit per package.