Learn More
Waters Corp Waters Corporation Tubing Assembly, CM-A Switching Valve to Detector, Low Pressure, MP35N, 14.5 in
Catalog No. 502039543 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement component designed for use in Bio ACQUITY systems. It connects the Column Manager-A Switching Valve to the detector, ensuring reliable low-pressure fluid transfer.
  • Constructed from MP35N.
  • Features low pressure fittings.
  • Measures 14.5 in long.
  • Used in Bio ACQUITY systems.
Catalog No. 50-203-9543 Supplier Waters Corp Supplier No. 700005482
This tubing assembly is a replacement component designed for use in Bio ACQUITY systems. It connects the Column Manager-A Switching Valve to the detector, ensuring reliable low-pressure fluid transfer.
  • Constructed from MP35N.
  • Features low pressure fittings.
  • Measures 14.5 in long.
  • Used in Bio ACQUITY systems.