Learn More
Waters Corp Waters Corporation Tubing, 1/16 in OD, 0.040 in ID
Catalog No. 502392713 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for connecting the degasser port A1 to the solvent select valve A. It features a 1/16 in outer diameter and a 0.040 in inner diameter, making it suitable for specific chromatographic applications.
  • Designed for connecting degasser port A1 to solvent select valve A.
  • Features a 1/16 in outer diameter.
  • Features a 0.040 in inner diameter.
Catalog No. 50-239-2713 Supplier Waters Corp Supplier No. 430001115
This tubing assembly is designed for connecting the degasser port A1 to the solvent select valve A. It features a 1/16 in outer diameter and a 0.040 in inner diameter, making it suitable for specific chromatographic applications.
  • Designed for connecting degasser port A1 to solvent select valve A.
  • Features a 1/16 in outer diameter.
  • Features a 0.040 in inner diameter.