Learn More
Waters Corp Tubing Assembly, GPV-A to Mixer, MP35N
Catalog No. 502390864 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a replacement component designed for the Bio Quaternary Solvent Manager GPV-A to Mixer. It is constructed from MP35N and is specifically used in H-Class Bio ACQUITY systems.
  • Replacement tube assembly for GPV-A to Mixer.
  • Constructed from MP35N.
  • Compatible with H-Class Bio ACQUITY systems.
  • Supports ACQUITY H-Class PLUS Bio Quaternary Solvent Manager.
  • Supports ACQUITY UPLC H-Class Bio QSM.
Catalog No. 50-239-0864 Supplier Waters Corp Supplier No. 430002481
This tubing assembly is a replacement component designed for the Bio Quaternary Solvent Manager GPV-A to Mixer. It is constructed from MP35N and is specifically used in H-Class Bio ACQUITY systems.
  • Replacement tube assembly for GPV-A to Mixer.
  • Constructed from MP35N.
  • Compatible with H-Class Bio ACQUITY systems.
  • Supports ACQUITY H-Class PLUS Bio Quaternary Solvent Manager.
  • Supports ACQUITY UPLC H-Class Bio QSM.