Learn More
Waters Corp Tubing Assembly, High Pressure, 0.003 in. SS, 14.5 in.
Catalog No. 502391982 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for high-pressure applications. It features a welded stainless steel construction with specific dimensions for reliable performance.
  • Welded tube assembly
  • Constructed from stainless steel
  • 0.003 in. ID
  • 14.5 in. long
Catalog No. 50-239-1982 Supplier Waters Corp Supplier No. 700008869
This tubing assembly is designed for high-pressure applications. It features a welded stainless steel construction with specific dimensions for reliable performance.
  • Welded tube assembly
  • Constructed from stainless steel
  • 0.003 in. ID
  • 14.5 in. long