Learn More
Waters Corp WATERS CORPORATION Tubing Assembly, Low Pressure, MP35N, 0.004 x 36 in.
Catalog No. 502039625 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is an ACQUITY H-Class Bio Welded Tube Assembly, constructed from MP35N material. It is designed for low pressure applications with dimensions of 0.004 x 36 in., and serves as a crucial spare part for compatible systems.
  • ACQUITY H-Class Bio Welded Tube Assembly
  • Made from MP35N material
  • Measures 0.004 in. x 36 in.
  • Designed for low pressure applications
  • Supplied as 1 unit per package
  • Replacement is recommended as required
Catalog No. 50-203-9625 Supplier Waters Corp Supplier No. 700008915
This tubing assembly is an ACQUITY H-Class Bio Welded Tube Assembly, constructed from MP35N material. It is designed for low pressure applications with dimensions of 0.004 x 36 in., and serves as a crucial spare part for compatible systems.
  • ACQUITY H-Class Bio Welded Tube Assembly
  • Made from MP35N material
  • Measures 0.004 in. x 36 in.
  • Designed for low pressure applications
  • Supplied as 1 unit per package
  • Replacement is recommended as required