Learn More
Waters Corp Tubing Assembly, Mixer Manifold to i2V, MP35N
Catalog No. 502390761 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tube assembly connects the Quaternary Solvent Manager Mixer Manifold to the i2V check valve. It is constructed from MP35N and is used in H-Class Bio ACQUITY systems.
  • Replacement tube assembly
  • Connects Quaternary Solvent Manager Mixer Manifold to i2V check valve
  • Constructed from MP35N
  • Used in H-Class Bio ACQUITY systems
Catalog No. 50-239-0761 Supplier Waters Corp Supplier No. 430002479
This replacement tube assembly connects the Quaternary Solvent Manager Mixer Manifold to the i2V check valve. It is constructed from MP35N and is used in H-Class Bio ACQUITY systems.
  • Replacement tube assembly
  • Connects Quaternary Solvent Manager Mixer Manifold to i2V check valve
  • Constructed from MP35N
  • Used in H-Class Bio ACQUITY systems